Puerto Rico
Order Entry
Anti-CD11c Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 3.9]
Anti-CD11c Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 3.9]
Supplier:  Biolegend
Certificates
Anti-CD11c Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 3.9]
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 301625-BL
  • 301626-BL
  • Antigen Symbol:
  • CD11c
  • CD11c
  • Antigen Synonyms:
  • aX integrin|Integrin αX subunit|ITGAX|CR4|p150
  • aX integrin|Integrin αX subunit|ITGAX|CR4|p150
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PB (Pacific Blue)
  • PB (Pacific Blue)
  • Clone:
  • 3.9
  • 3.9
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3687
  • 3687
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
  • The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Product Information

  • Catalog #
    Supplier #
    Size
    Supplier no.
    Availability
    Price
  • 301625-BL
    301625
    25 tests
    301625
    Log in for availability
    Log in for availability
    Each
    Each - $155.50
    $155.50
    Each
       $155.50

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Antigen Symbol
    CD11c
    Environmentally Preferable
    Antigen Synonyms
    aX integrin|Integrin αX subunit|ITGAX|CR4|p150
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PB (Pacific Blue)
    Clone
    3.9
    Reactivity
    Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
    Host
    Mouse
    Gene ID
    3687
    Isotype
    IgG1 kappa
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
    Purification
    The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT
  • 301626-BL
    301626
    100 tests
    301626
    Log in for availability
    Log in for availability
    Each
    Each - $353.90
    $353.90
    Each
       $353.90

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Antigen Symbol
    CD11c
    Environmentally Preferable
    Antigen Synonyms
    aX integrin|Integrin αX subunit|ITGAX|CR4|p150
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PB (Pacific Blue)
    Clone
    3.9
    Reactivity
    Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
    Host
    Mouse
    Gene ID
    3687
    Isotype
    IgG1 kappa
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
    Purification
    The antibody was purified by affinity chromatography, and conjugated with Pacific Blue™ under optimal conditions. The solution is free of unconjugated Pacific Blue™ .
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT