Puerto Rico
Order Entry
Anti-Biotin Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1D4-C5]
Anti-Biotin Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1D4-C5]
 :  Biolegend
 
Anti-Biotin Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1D4-C5]
 :  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 409004-BL
  • 409003-BL
  • Antigen Symbol:
  • Biotin
  • Biotin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 1D4-C5
  • 1D4-C5
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • Biotinylated limulus hemocyanin
  • Biotinylated limulus hemocyanin
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

 

  •  
     
    Size
    Supplier no.
     
     
  • 409004-BL
    409004
    100 μg
    409004
       
       
    Each (100µG)
    Each (100µG) - $315.10
    $315.10
    Each (100µG)
       $315.10

     
     
     
    Specifications
    Antigen Symbol
    Biotin
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PE (Phycoerythrin)
    Clone
    1D4-C5
    Reactivity
    Host
    Mouse
    Isotype
    IgG2a kappa
    Immunogen
    Biotinylated limulus hemocyanin
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
    Purification
    The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
    Concentration
    0.20 mg/mL
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT
  • 409003-BL
    409003
    25 μg
    409003
       
       
    Each (25µG)
    Each (25µG) - $90.10
    $90.10
    Each (25µG)
       $90.10

     
     
     
    Specifications
    Antigen Symbol
    Biotin
    Environmentally Preferable
    Antibody Type
    Primary
    Clonality
    Monoclonal
    Conjugation
    PE (Phycoerythrin)
    Clone
    1D4-C5
    Reactivity
    Host
    Mouse
    Isotype
    IgG2a kappa
    Immunogen
    Biotinylated limulus hemocyanin
    Form
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
    Purification
    The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
    Concentration
    0.20 mg/mL
    Storage Temperature
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
    Shipping Temperature
    Ambient RT