BRAND® Dispensette® S and Dispensette® S Organic Bottletop Dispensers, BrandTech®
Supplier: BrandTech
4600330 4600310 4600210 4600121 4630341 4630160 4600161 4630170 4630230 4630140 4630360 4600331 4630330 4600130 4600220 4600351 4630151 4600170 4600231 4630331 4600360 4600320 4600340 4600341 4600361 4630161 4600151 4630351 4600321 4630240 4600211 4600160 4630241 4600171 4630131 4600230 4600150 4630361 4600240 4600100 4630171 4600311 4630340 4600140 4600241 4600221 4600131 4600350 4630141 4630130 4630350 4600120 4600101 4630231 4630150 4600141
10018-966EA 731.2 USD
10018-966 10018-968 10018-962 10018-964 10019-010 10019-014 10019-012 10019-006 10019-004 10019-008 10018-970 10018-972 10018-934 10018-978 10018-936 10018-974 10018-930 10018-976 10018-932 10019-020 10019-024 10018-938 10019-022 10019-018 10019-016 10018-980 10018-982 10018-944 10018-988 10018-946 10018-984 10018-940 10018-986 10018-942 10018-948 10020-964 10020-966 10020-968 10018-992 10018-950 10018-994 10020-960 10020-962 10018-990 10018-956 10018-958 10018-996 10018-952 10018-954 10018-998 10019-002 10019-000 10020-974 10018-960 10020-970 10020-972
BRAND® Dispensette® S and Dispensette® S Organic Bottletop Dispensers, BrandTech®
Bottle-Top Dispensers
The new Dispensette® S builds on a half century of BRAND® bottletop dispenser experience. It features a floating piston design with even lower operating forces, a new safety discharge system, and an improved sealing system, eliminating sealing washers for even better reliability. Designed to attach directly to a reagent bottle, eliminating poured transfer. In addition to the product improvements, a new 0.1–1mL size is available.
  • Floating piston eliminates seal wear
  • Safety discharge system
  • Better reliability
  • Fits most reagent bottles directly
Choose the Dispensette® S for general-purpose dispensing of acids, bases, buffers, and many common solvents (NOT suitable for HF)
Choose from two different volume adjust mechanisms or fixed volume models. Analog adjustable models feature a sliding pointer with scalloped inner track, well suited for rapid volume changes from one end of the range to the other. Digital adjustable models are excellent for highly reproducible volume settings. All are available with or without a recirculation valve that allows easy priming without ‘spitting’ or reagent wasting. Many accessories, including adapters for nearly any unpressurized container, are available.
For hydrofluoric acid, choose the Dispensette® S Trace Analysis platinum-iridium. For concentrated acids, TFA, peroxides, THF, and other difficult solvents, choose the Dispensette® S Organic. A selection chart is available.
Información accesorios: Discharge tubes are available separately as accessories. Tubes feature threaded safety caps, and are color coded to match the appropriate Dispensette®. Adjustable length filling tubes require no cutting. Discharge tubes come in standard and fine oricifice tips. Flexible discharge tubes feature 800mm of coiled PTFE tubing with a weighted, textured safety grip.
Información para pedidos: Dispensette® S and Dispensette® S Organic bottle-top dispensers are supplied with performance certificate, telescoping filling tube, recirculation valve and recirculation tube (if specified), valve mounting tool, and a selection of polypropylene adapters. Bottle not included.
Precaución: Flexible discharge tubes must never be used with hydrofluoric acid (HF).