Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • SBE (20)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

PCR Buffers

https://pr.vwr.com/stibo/smallweb/std.lang.all/69/31/20236931.jpg

PCR buffer solutions provide a suitable chemical environment for optimum activity and stability of the DNA polymerase during polymerase chain reactions. The usage of alternate buffer components or polymerase enzymes can help with amplification of long or otherwise problematic regions of DNA. The addition of reagents in buffer systems may increase the specificity and yield of PCR. PCR buffer is necessary to create optimal conditions for activity of Taq DNA polymerase.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"27"
Sort by:
 
 
 
 

Supplier:  Lucigen
Description:   Highly specific thermostable DNA ligase for high-temperature applications
Supplier:  Lucigen
Description:   Purchase individual FailSafe™ DNA polymerase and/or FailSafe™ 2X PreMix Buffers here
Supplier:  New England Biolabs (NEB)
Description:   OneTaq® Quick-Load® 2X Master Mix with Standard Buffer is an optimized, ready-to-use blend of Taq and Deep VentR ™ DNA Polymerases ideally suited to r...
Small Business Enterprise
Supplier:  Zymo Research
Description:   Sequencing Wash Buffer, Size: 500 ml^
Supplier:  New England Biolabs (NEB)
Description:   OneTaq® 2X Master Mix with Standard Buffer is an optimized blend of Taq and Deep VentR™ DNA Polymerases ideally suited to routine PCR applications fro...
Small Business Enterprise
Supplier:  Promega Corporation
Description:   5X Green and 5X Colorless GoTaq Flexi Reaction Buffers are Mg-free buffers that allow optimization of Mg concentration in PCR. The green buffer allows...
SDS
Catalog Number: (PAP1121)
Supplier:  Promega Corporation
Description:   Riboprobe System Buffers are qualified for use in applications where maintaining the integrity of RNA is critical. Product is quality tested to ensure...
Supplier:  New England Biolabs (NEB)
Description:   OneTaq® Hot Start Quick-Load® 2X Master Mix with GC Buffer is an optimized, ready-to-use blend of Taq and Deep VentR™ DNA Polymerases combined with an...
Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 27
  1  2  3  4  Next