CryoPod Carrier provides safe, reliable and portable <-150˚C cryogenic transport for biospecimens for over 4 hours
- Operator safety
- Over four hours hold time
- Maintains sample integrity
- Reliable performance
- Compact, lightweight
The CryoPod Carrier allows safe and quick transportation of cryogenic samples and features an auto-fill option. The entire working depth remains well below -150˚C for over 4 hours with one charge of LN2. The carrier features a temperature display with audible and visual alarms, a megnetized lid and built-in handle and finger grips. The unit accomodates one standard freezer storage box. There is no direct sample contact with LN2. The CryoPod Carrier allows for temperature logging and retrieval, and it requires only 3 Liters of liquid nitrogen.
Use around the laboratory, campus or short transport between facilities. Displays temperature, date and time, and features audible and visual alarms, as well as logging capability. Integrates into an (optional) automated filling station or can be manually filled using the included fill kit.
Información para pedidos: CryoPod includes the unit, one CryoPod foam lid insert in green, a manual fill kit, batteries for display and a pack of LN2 foam absorbent pads. Additional foam pads are available in pack of 4. Lid inserts are available in 5 colors.