BD INTRAMEDIC™ Polyethylene Tubing, Sterile, BD Medical
Supplier: BD Medical
427516 427517 427517
20903-414PK 535.06 USD
CA63020-616 CA20903-414 20903-414
BD INTRAMEDIC™ Polyethylene Tubing, Sterile, BD Medical
Tubing Transfer Tubing
Polyethylene tubing is flexible, but rigid enough for applications using vacuum and pressure.
  • Low water absorption rate
  • Waxy white, translucent, tasteless, odorless
  • Sterile, non-pyrogenic, and non-radiopaque
  • Polyethylene contains no additives or plasticizers
Tubing is resistant to most common acids and bases. Durable over a wide temperature range, up to 104 °C (219 °F).