Sintered Glass Fiber Filter Discs, Porosity D, Ace Glass Incorporated
Supplier: Ace Glass
7176-125 7176-135 7176-30 7176-35 7176-60 7176-15 7176-04 7176-40 7176-70 7176-80 7176-145 7176-165 7176-103 7176-45 7176-50 7176-25 7176-20 7176-09 7176-155 7176-115 7176-55
89057-894EA 33.93 USD
89057-894 89057-982 89057-992 89057-788 89057-942 89057-766 89057-952 89057-776 89057-962 89057-884 89057-972 89057-836 89057-846 89057-824 89057-746 89057-812 89057-800 89057-932 89057-756 89057-816 89058-002
Sintered Glass Fiber Filter Discs, Porosity D, Ace Glass Incorporated
Filters Glass Fiber Filters
These glass fiber filter discs last longer and are more abrasion resistant.
  • Superior in performance to granular type discs
  • Resistant to thermal shock and chemical attack
  • Fabricated from borosilicate glass
Available in a complete range of porosities and diameters, these discs can be used in the manufacture of a wide variety of special apparatus.