Sintered Glass Fiber Filter Discs, Porosity B, Ace Glass Incorporated
Supplier: Ace Glass
7176-143 7176-07 7176-43 7176-101 7176-68 7176-53 7176-38 7176-02 7176-123 7176-153 7176-48 7176-18 7176-33 7176-13 7176-78 7176-58 7176-163 7176-23 7176-133 7176-28 7176-113
89057-890EA 33.04 USD
89057-890 89057-762 89057-784 89057-872 89057-772 89057-792 89057-832 89057-820 89057-842 89057-742 89057-752 89057-796 89057-968 89057-978 89057-988 89057-998 89057-938 89057-948 89057-958 89057-808 89057-918
Sintered Glass Fiber Filter Discs, Porosity B, Ace Glass Incorporated
Filters Glass Fiber Filters
These glass fiber filter discs last longer and are more abrasion resistant.
  • Superior in performance to granular type discs
  • Resistance to thermal shock and chemical attack
  • Fabricated from borosilicate glass
Available in a complete range of porosities and diameters, these discs can be used in the manufacture of a wide variety of special apparatus.