Sintered Glass Fiber Filter Discs, Porosity A, Ace Glass Incorporated
Supplier: Ace Glass
7176-142 7176-100 7176-162 7176-27 7176-22 7176-152 7176-12 7176-06 7176-132 7176-42 7176-17 7176-47 7176-57 7176-112 7176-67 7176-32 7176-52 7176-01 7176-37 7176-122 7176-77
89057-780EA 25.79 USD
89057-780 89057-740 89057-750 89057-794 89057-760 89057-782 89057-770 89057-986 89057-864 89057-996 89057-830 89057-840 89057-946 89057-956 89057-966 89057-888 89057-976 89057-910 89057-818 89057-806 89057-936
Sintered Glass Fiber Filter Discs, Porosity A, Ace Glass Incorporated
Filters Glass Fiber Filters
These glass fiber filter discs last longer and are abrasion resistant.
  • Superior in performance to granular type discs
  • Available in a complete range of porosities and diameters, these discs can be used in the manufacture of a wide variety of special apparatus
  • Fabricated from borosilicate glass