Corning® 96-Well, High Binding, Half Area Microplate, Corning
Supplier: Corning
Corning®
7442
10215-520CS 2760.53 USD
10215-520
Corning® 96-Well, High Binding, Half Area Microplate, Corning
Microplates Cell Culture Plates
These Corning® Half Area Microplates have a reduced well volume of 205μL
The optically clear flat bottom permits direct micropscope viewing. Opaque walls are designed to prevent well-to-well cross talk.