Puerto Rico
Order Entry
Strontium nitrate ≥99.0% ACS
Strontium nitrate ≥99.0% ACS
 
Strontium nitrate ≥99.0% ACS

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Assay:
    99.0% min.
  • pH of 5% solution at 25°C:
    5.0-7.0
  • Insoluble matter:
    ≤0.01%
  • Loss on drying at 105°C:
    ≤0.1%
  • Chloride (Cl):
    ≤0.002%
  • Sulfate (SO4):
    ≤0.005%
  • Barium (Ba):
    ≤0.05%
  • Calcium (Ca):
    ≤0.05%
  • Magnesium (Mg):
    ≤0.10%
  • Sodium (Na):
    ≤0.10%
  • Heavy metals (as Pb):
    ≤5ppm
  • Iron (Fe):
    ≤5ppm

 

 

 

  •  
     
    Size
    Supplier no.
     
     
  • BT142365-25G
    142365-25G
    25 g
    142365-25G
       
       
    Each (25g)
    Each (25g) - $35.17
    $35.17
    Each (25g)
       $35.17

  • BT142365-250G
    142365-250G
    250 g
    142365-250G
       
       
    Each (250g)
    Each (250g) - $54.29
    $54.29
    Each (250g)
       $54.29

  • BT142365-1KG
    142365-1KG
    1 kg
    142365-1KG
       
       
    Each (1kg)
    Each (1kg) - $152.34
    $152.34
    Each (1kg)
       $152.34