Gosselin™ Conical Containers with Snap Cap, 400 ml, Corning®
Supplier: Corning Life Sciences
PC400-09
89238-028CS 99.89 USD
89238-028
Gosselin™ Conical Containers with Snap Cap, 400 ml, Corning®
Sample Containers General Purpose Containers
Gosselin™ Conical Containers are supplied with snap cap and are designed to collect and store samples before testing
These nonsterile polypropylene containers offer a 400mL capacity, and feature graduations.