Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
: Biotium
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 71004-192
- 89138-386
- Antigen Symbol:
- IgG
- IgG
- Antigen Name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Reactivity:
- Rabbit
- Rabbit
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Epitope:
- Heavy and Light chain
- Heavy and Light chain
- SizeSupplier no.
Specifications
Antigen SymbolIgGAntigen NameImmunoglobulin GEnvironmentally PreferableAntibody TypeSecondaryClonalityPolyclonalConjugationCF® 488AReactivityRabbitHostGoatIsotypeIgGWestern BlotYesELISAYesImmunoFluorescenceYesFlow CytometryYesEpitopeHeavy and Light chainSpecifications
Antigen SymbolIgGAntigen NameImmunoglobulin GEnvironmentally PreferableAntibody TypeSecondaryClonalityPolyclonalConjugationCF® 488AReactivityRabbitHostGoatIsotypeIgGWestern BlotYesELISAYesImmunoFluorescenceYesFlow CytometryYesEpitopeHeavy and Light chain