To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Adipocyte Plasma Membrane Associated Protein
- Antigen Symbol:APMAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Epitope:Maps to amino acids 230-244 of Human Adiponectin protein
- Cross Adsorption:No
- Format:Liquid
- Antigen Synonyms:a|Adipose most abundant gene transcript 1|30 kDa adipocyte complement-related protein|Adipocyte|ACRP30|C1q and collagen domain-containing protein
- Immunogen:A synthetic peptide from human Adiponectin (230-244 aa).
- Cat. No.:10781-946
- Supplier no.:C-1524-100
Adiponectin is synthesized by adipocytes and is involved in the control of fat metabolism and insulin sensitivity, with direct anti-diabetic, anti-atherogenic and anti-inflammatory activities (ref: SWISSPROT).
This antibody detects adiponectin protein at approx 30 kDa in western blots from human, mouse and rat samples. Plasma makes a good positive control. The antibody has not been tested in IHC
Application Information:
WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: APMAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 230-244 of Human Adiponectin protein
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat