Anti-LAT2 Mouse Monoclonal Antibody [clone: NAP-07]
10723-312
: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Linker For Activation Of T Cells Family, Member 2
- Antigen Symbol:LAT2
- Clonality:Monoclonal
- Clone:NAP-07
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:7462
- Antigen Synonyms:WBSCR5|Williams-Beuren syndrome chromosomal region 15 protein|membrane-associated adapter molecule|transmembrane adaptor 2|HSPC046|non-T-cell activation linker|linker for activation of T-cells family member 2|WBSCR15|WSCR5|LAB|Williams-Beuren syndrome chromosomal region 5 protein|linker for activation of B-cells|linker for activation of T cells
- Amino Acid Number:91 to 243
- Storage Buffer:PBS, pH 7.4 (15 mM sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 91-243 of human LAT2.
- Cat. No.:10723-312
- Supplier no.:MAB0946
Mouse monoclonal antibody raised against partial recombinant LAT2.
Type: Primary
Antigen: LAT2
Clonality: Monoclonal
Clone: NAP-07
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse