Anti-mCherry Polyclonal Antibody
10725-114
: Avantor
- Antibody Type:Primary
- Antigen Name:mCherry
- Antigen Symbol:mCherry
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal)
- Storage Temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to mCherry.
- Cat. No.:10725-114
- Supplier no.:PAB18013
Rabbit polyclonal antibody raised against recombinant mCherry.
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml
Type: Primary
Antigen: mCherry
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: