Anti-MST1R Mouse Monoclonal Antibody [clone: 1H7]
10593-930
: Abnova
- Antibody Type:Primary
- Antigen Name:Macrophage Stimulating 1 Receptor
- Antigen Symbol:MST1R
- Clonality:Monoclonal
- Clone:1H7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Size:100 µg
- Cross Adsorption:No
- Gene ID:4486
- Antigen Synonyms:soluble R|RON variant E2E3|MST1R variant RON30|PTK8|macrophage-stimulating protein receptor|MST1R variant RON62|soluble RON variant 1|c-met-related tyrosine kinase|CDw136|PTK8 protein tyrosine kinase 8|MSP receptor|p185-Ron|soluble RON variant 3|RON|soluble RON variant 2|CD136
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10593-930
- Supplier no.:H00004486-M07
Type: Primary
Antigen: MST1R
Clonality: Monoclonal
Clone: 1H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human