Would you like to log in again?
Velcro Cup Heel Grounder, Transforming Technologies
Supplier: TRANSFORMING TECHNOLOGIES
HG1594
14224-328CS 1677.71 USD InStock
14224-328
Velcro Cup Heel Grounder, Transforming Technologies
Grounders Heel and Toe Grounders
The Transforming Technologies velcro cup heel grounder is constructed from durable materials and features a black stretch loop closure to maximize fit and comfort
  • Stretch loop closure
  • Buried resistor 1 meg ohm (+/- 5% tolerance)
  • A \<10e5 ohms, 1.25" wide rubber exterior
  • Conductive yarn in gray polyester ribbon
  • Available in black, red, blue or orange
These heel grounders connect the person wearing them to ground via a proper floor mat or flooring material. Wearing the conductive ribbon inside the shoe or sock ensures proper electrical contact with the user. The ribbon connects to the conductive rubber to complete the circuit between the operator and the static controlled floor or mat. A buried 1 meg ohm resistor is standard. These heel grounders are worn on both feet to provide consistent grounding while in motion.
Certificaciones: Meets or exceeds requirements of ANSI ESD-S20.20.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty