Splash Guard, Clear Acrylic, TrippNT
Supplier: TrippNT
50047
10012-670EA 778.49 USD
10012-670
Splash Guard, Clear Acrylic, TrippNT
Shields Safety Shields
Beef up protection when working with caustic or hot materials with this acrylic splash guard. The splash guard provides a cost-effective and efficient way to protect your face, chest and arms from splashing liquids, samples or reagents.
  • Heavy duty 3/8" acrylic to protect from caustic or corrosive liquids
  • Completely transparent from all angles
  • Defined side shields
  • Extra sturdy and tough
  • In stock and ready to ship
  • American made by TrippNT
  • Total replacement Lifetime guarantee
Constructed with thick, completely see-through acrylic for added strength and uncompromisable visibility. 3/8" acrylic construction, curved sides, unrestricted access and visibility when working with materials that may splash or injure.