Anti-Kallikrein 1 Rabbit Polyclonal Antibody
10209-262
- Antibody Type:Primary
- Antigen Name:Kallikrein 1
- Antigen Symbol:KLK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Form:Lyophilized
- Amino Acid Number:25 to 261
- Storage Temperature:–20 °C
- Immunogen:E. coli-derived mouse KLK1 recombinant protein (Position: I25-D261).
- Purification:purified by immunogen affinity purification
- Cat. No.:10209-262
- Supplier no.:PA2038
Rabbit IgG polyclonal antibody for Kallikrein-1(KLK1) detection. Tested with WB, IHC-P in Mouse.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Kallikrein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse