Puerto Rico
Order Entry
Human CXCL10/IP-10 PicoKine ELISA Kit, Boster
Catalog # 10205-298
CAS Number:  
undefined
Human CXCL10/IP-10 PicoKine ELISA Kit, Boster
  10205-298
 :  EK0735
 :  
Price
$1,230.59
Log in for availability
Unit of Measure Each
Minimum order quantity is 1
 

 

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    IP-10 (CXCL10)
  • Size:
    96 well
  • Immunogen:
    Standard: E.coli, V22-P98
  • Cat. No.:
    10205-298
  • Supplier no.:
    EK0735