SHURMount II Automated Glass Coverslipper, Triangle Biomedical
SMAGC-II
10062-794EA 66413.25 USD
10062-794
SHURMount II Automated Glass Coverslipper, Triangle Biomedical
The SHURMount II Automated Glass Coverslipper delivers the reliable speed and throughput  your busy lab demands to achieve highly accurate results.  Loading capacity 60 slides; Coverslipping speed of 14 sec/slide
  • Compatibility with a variety of automated stainers
  • Intuitive user interface for Easy access to essential functions
  • Exclusive dispensing system provides bubble-free coverslipping
  • Compact footprint requires minimal counter space
  • Reliable speed and accuracy for workflow efficiency
Utilizing the SHURMount™ Automated Glass Coverslipper with General Data’s broad histology product line of instrumentation, consumable & reagents we’re your one source for all your laboratory needs.
Its compact footprint is ideal for crowded workspaces that gives labs maximum for instrument layout and work-flow design. The versatile functionality allows it to operate as a stand-alone unit or in combination with a variety of other automated stainers. Its user-friendly control panel provides complete control over all coverslipping functions.
Información para pedidos: Includes 3 each Slide Baskets, Storage/Drying Racks and Universal Coverslip Hopper.