Corepoint Scientific™ Refrigerated Incubators, Stability Chamber, Horizon Scientific
Supplier: Horizon Scientific
NSRI522WSW/0
10714-180EA 21674.99 USD
10714-180
Corepoint Scientific™ Refrigerated Incubators, Stability Chamber, Horizon Scientific
Corepoint Scientific™ Stability Chamber-Refrigerated Incubator, Digital 132 x 64 LED display, Programmable logic (PLC) microprocessor temperature controller, Audible and visual Hi/Lo temperature alarm, Adjustable control range from +4°C to +70°C, Programmable ramp and soak, Remote alarm contacts, Continuous product temperature display,3 sensor system; 1 product, 1 air, 1 coil, Sensor access port 1" diameter, Power supply breaker and convenience power switch located on facade, Door ajar alarm, audible and visual, Temperature transmitter (0-10 VDC) (selectable to air or product), RS-485 digital communications port (MODBUS-RTU), Password protection - set-points and factory settings, 100 event alarm logging date/time stamp, Integrated Hi/Lo product temperature alarm test, Digital calibration air and product, Real time clock with date and time display.
Door opening counter with 24 hour automatic reset, Adjustable alarm beep or constant on (key pad selectable), Low battery test switch with buzzer, Perimeter anti-condensate door frame heater (on/off temperature selectable), Sensor failure alarm, Power failure alarm, Exterior cabinet front, sides and back are powder coated, Exterior cabinet top and bottom are galvanized steel, Interior cabinet liner is stainless steel, Cabinet is foamed-in-place with CFC-free high density, polyurethane foam insulation, Solid door(s), full size, Heavy duty door pivot hinges, Pull door handles, full length, Magnetic door gaskets, Air cooled condensing unit, Centered key door lock(s) (2 keys), Four casters (2 locking) on one and two door models, six, casters (3 locking) on three door models, 3 coated wire shelves per door section, Top mounted forced air refrigeration system, Condensate evaporation, LED lighting.
Exterior front, sides and back powder coated. Exterior top and bottom glavanized steel. Interior cabinet is stainless steel.
Certificaciones: UL, C-UL listed (60 Hz models only).
Información para pedidos: Accessories not included. All Units include 18 months limited warranty on parts and labor (US & Canada). Warranties include 18 months parts and labor, 5 year compressor, (US and Canada), 18 months parts (International).