87-276 gloves feature a unique 20-millimeter neoprene and natural rubber latex blend and provide excellent chemical, snag, puncture, and abrasion resistance.
- Extra grip on wet or dry surfaces
- Excellent resistance to chemicals, punctures, snags, and abrasions
- Useful in a range of industries
Pebble embossing provides added grip on wet or dry surfaces.
Use in the aerospace, agriculture and viticulture, automotive and transportation, chemical, food handling and processing, general handling, and petrochemical industries. Uses include canning and food processing; chemical processing; dairy processing; fruit and vegetable processing; handling acids, alkalies, salts, ketones and alcohols; handling specialty chemicals; laboratory testing; meat and poultry processing; and sanitation and dish washing.
The 100% cotton flock lining offers increased comfort.
Certificaciones: Complies with FDA food handling requirements (21 CFR 177.2600). ANSI level 3 abrasion protection.
Envase: Gloves are packaged 12 pairs in a bag; 144 in a carton.
Precaución: This product contains natural rubber latex, which may cause allergic reactions in some individuals.