StoneScreen™ Kidney Stone Collection and Transport Kit, Therapak®
Supplier: Therapak
StoneScreen™
38879G
10497-206CS 184.7 USD
10497-206
StoneScreen™ Kidney Stone Collection and Transport Kit, Therapak®
Sample Containers Urine Containers
The StoneScreen™ collection and transport kit provides a means of collecting and transporting kidney stone samples for laboratory analysis
The kit includes the StoneScreen™ kidney stone collection cup, which is constructed to capture renal calculi, and designed for use with patients attempting to pass stones following extracorporeal shock wave lithotripsy. The urine-straining device is a collapsible plastic strainer cup designed for both men and women. It includes a nylon screen at the bottom of the cup for stone capture. Once the stone is captured, it can be placed in the storage space under the lid and saved for analysis. After the entire cup and sample is re-collapsed following collection, place the specimen cup into the zip style bag and return the specimen cup into the transport box along with the test requisition for courier pick up.