Gloves incorporate Ansell Grip™ technology for a revolutionary combination of excellent grip and strong oil repellence, and microscopic channels in glove coating guide away oil, resulting in secure glove-to-surface contact.
- Strong oil repellence
- Excellent grip with microscopic oil-wicking channels
- Secure glove-to-surface contact
- Users benefit from comfort, dryness, and superior tactility
This allows the wearer to confidently handle small- and medium-sized components lightly coated with oil or lubricant.
Use in the automotive and transportation, building and construction, machinery and equipment, maintenance, and metal fabrication industries. Uses include assembly and finishing; assembly of white goods; general site and factory maintenance; light metal fabrication and handling; metal fabrication; and quality control.
Gloves protect skin from oil contamination and incorporate all the essential features of the Ansell HyFlex range to provide users with fit, comfort, and dryness, as well as excellent dexterity and tactility.
Certificaciones: ANSI Level 1 Cut; ANSI Level 4 Abrasion.
Envase: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.