Dissolve 10 mg hyaluronic acid in 25 mL 0.1 M sodium phosphate buffer: pH 5.3 with 0.15 M sodium chloride.
Amount of enzyme which liberates one micromole of N-acetylglucosamine per minute at 37°C and pH 4.0
Inhibitors: Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+, Stabilizers: Sodium chloride acts as a stabilizer.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- CAS No.:37326-33-3
- Form:Lyophilized
- Enzyme Name:Hyaluronidase
- SizeSupplier no.