Feline Serum, ImmunoReagents
Supplier: Immunoreagents
IR2428 IR2429
10152-206EA 210.68 USD
10152-206 10152-208
Feline Serum, ImmunoReagents
Sera (Serum)
ImmunoReagents’ normal feline serum is used as a blocking agent or as a control for immunoassays
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.
All serum is obtained from healthy, disease free animals. Serum is lipid extracted and buffer exchanged against phosphate buffer. Our serum is supplied in lyophilized form without preservative. It is recommended to reconstitute the normal serum with DI water and add a preservative of choice, or to aliquot reconstituted serum and store frozen to prevent contamination.
Certificaciones: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Información para pedidos: ImmunoReagents products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles. Additionally, products can by customized based on specific assay requirements. Contact your VWR representative for more information.