Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LEUCOGNOST® PAS stain for hematology, Sigma-Aldrich®

Supplier: MilliporeSigma

Danger

1.16302.0002 1.16302.0002
EM1.16302.0002CS 1938.27 USD InStock
CA1.16302.0002 EM1.16302.0002
LEUCOGNOST® PAS stain for hematology, Sigma-Aldrich®
LEUCOGNOST®
Detection of the Periodic Acid Schiff reaction in leukocytes

The LEUCOGNOST PAS reaction is an important method for the identification of lymphatic cell elements. Next to peroxidase and esterase reactions, it is one of the three basic cytochemical staining methods important for differential diagnosis that are regularly carried out in acute cases of leukaemia. Smears already stained by the Pappenheim method can additionally be stained with PAS and the stains subsequently removed with 1% periodic acid. It is developed and registered as CE / IVD product for applications in diagnostics.The staining kit is suffi cient for 12 stainings with up to 16 slides.


Density: 0.82 g/cm³ (20 °C)
Storage Temperature: Ambient
EINECS: 000-000-0
UN: 3085
ADR: 5.1 (8),I
http://media.vwr.com//stibo/web/std.lang.all/23/03/52303.jpg
Order Now
Specification Test Results
Suitability for microscopy (Blood smear) passes test
PAS positive substances red
Nuclei blue


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty