Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Chicken IgG Antibody (AzureSpectra 700)

Supplier: Azure Biosystems

AC2131
10147-356EA 97.3 USD InStock
10147-356
Goat Anti-Chicken IgG Antibody (AzureSpectra 700)
Antibodies
Azure offers a wide range of secondary antibodies labeled with fluorophores detected in infrared and near-infrared wavelengths.

AzureSpectra 700 and 800 secondary antibodies deliver enhanced sensitivity relative to visible fluorescent Western experiments because of low background autofluorescence in the near-infrared region.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: AzureSpectra 700
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty