Electroporation System, Twin Wave, Gemini X², X² HT and SC2, BTX
45-2007 45-2002 45-2007 45-2008 45-2002 45-2008
82428-570EA 19885.61 USD
82428-570 CA11032-098 82428-568 82428-566 CA11033-000 CA11033-002
Electroporation System, Twin Wave, Gemini X², X² HT and SC2, BTX
  • Simple user interface for easy touch screen programming
  • Resistance measurement helps to avoid arcing, which can damage cells; voltage accuracy ±5%
  • Multi-pulsing exponential decay allows for true high throughput 96-well pulsing for bacteria
  • Software for quick protocol updating and remote operations for robotics; software updates can be performed remotely
  • Log data storage for record keeping and troublehooting
The Gemini X² system is designed for researchers who need ultimate experiment flexibility in all cell types. In one easy setup, square wave and exponential decay waves can be applied to cells in any format. Electroporation of suspension cells can be achieved in cuvettes and 96 well plates. Additionally, the Gemini X² can be paired with BTX speciality electrodes to deliver genes and drugs in vivo, in utero, in ovo, to ex plant tissues as well as suspension cells. It incorporates remote operation functionality via footswitch or PC and internal log storage of experiment data for easy optimisation, quality control requirements and troubleshooting. It has 80 pre-set protocols with unlimited custom protocol storage.
The Gemini SC² system is essential for researchers electroporating cells in suspension. In one simple setup, square wave and exponential decay waves can be applied to cells in cuvettes. With a wide range of pulsing parameters, advanced safety features as well as dozens of pre-set protocols, the Gemini SC² can be used in any laboratory requiring efficient cell transfection or transformation without the use of costly reagents. It has 50 pre-set protocols for mammalian and bacterial applications with unlimited custom protocol storage.
Certificaciones: RoHS compliant
Información para pedidos: Complete Gemini X² system includes Gemini X² generator, safety dome, 30 cuvettes (10 each 1, 2 and 4 mm gap size), and cuvette rack. Complete Gemini X² HT system also includes HT 200 plate handler, 1×2 mm HT plate and 1×4 mm HT plate. Speciality electrodes are available separately. Complete Gemini SC² system includes Gemini SC² generator, safety dome, 30 cuvettes (10 each 1, 2 and 4 mm gap size), and cuvette rack.