MaxyClear™ Snaplock Microcentrifuge Tubes, 5 mL, Polypropylene, Axygen Scientific
Supplier: Corning
MaxyClear™
MCT-500-C-S MCT-500-C-S MCT-500-X MCT-500-C MCT-500-C MCT-500-C-S
89425-665CS 584.62 USD
89425-667 CA10159-772 89425-669 CA10159-802 89425-666 89425-665
MaxyClear™ Snaplock Microcentrifuge Tubes, 5 mL, Polypropylene, Axygen Scientific
Tubes Microcentrifuge Tubes
Snaplock microcentrifuge tubes 5 ml fit in standard 15 ml centrifuge rotors and offer the ultimate experience in lower volume sample requirements.
  • Smaller volume tubes minimize contamination issues
  • Pipet tips can easily reach into the tube for sample accessibility
  • Certified RNase-/DNase-free and non-pyrogenic for total sample integrity
These polypropylene microtubes feature frosted cap surfaces for labeling, frosted panels on the side for writing, and graduations at 0.5 ml increments.