i-RAMAN Plus Portable Fiber Optic RAMAN Systems, B&W Tek
840000401 840000304 810000389 840000139 840000381 840000387 840000395 840000009 840000287 840000327 840000217 840000321 810000503 840000360 810000468 840000346 810000589 840000388 840000323 840000324 840000101 840000363 840000127 840000340 810000501 840000328 840000325 840000126 840000400 810000044 840000117 840000136 840000326 840000218 840000322 840000131 840000129 810000502
10121-498EA 1513.83 USD
10121-498 10122-402 10122-400 10122-366 10122-388 10122-406 10122-328 10122-306 10122-404 10122-304 10122-382 10122-380 10122-320 10122-364 10122-386 10122-390 10121-466 10122-408 10122-308 10122-336 10122-314 10122-412 10122-334 10122-312 10122-378 10122-410 10122-318 10122-316 10122-372 10122-394 10122-370 10122-392 10122-376 10122-310 10122-398 10122-332 10122-374 10122-396
i-RAMAN Plus Portable Fiber Optic RAMAN Systems, B&W Tek
The i-RAMAN Plus is an enhanced version of our award winning i-RAMAN portable RAMAN spectrometer, now powered by our innovative smart spectrometer technology
The i-RAMAN Plus features the unique combination of wide spectral coverage and high resolution with configurations measuring out to 4000cm-1, enabling you to measure stretching bands around 3100cm-1. The system’s small footprint, lightweight design, and low power consumption provide research grade RAMAN capabilities anywhere. The i-RAMAN Plus comes standard with a fiber optic probe, and can be used with an XYZ positioning stage probe holder, a cuvette holder, and a trial version of our proprietary BWIQ multivariate analysis software. With the i-RAMAN Plus, a high precision qualitative and quantitative RAMAN solution is at your fingertips.
To enhance the functionality of the i-RAMAN Plus for user applications there is an extensive suite of software available at additional cost including: BWID Pharma Identification Software; BWIQ Multivariate Analysis software; BWSP 21pt11 Spectroscopy Analysis Software; and BWQT-M 21pt11 Chemometrics Model Builder. Functionality can be further enhanced with the addition of the optional BAC151B RAMAN Video Microsampling System, a powerful tool designed to offer the highest level of flexibility in facilitating Raman sampling for a variety of applications. Other optional accessories include a RAMAN probe holder BAC150B and a Raman cuvette holder BCR100A.
Using a high quantum efficiency CCD array detector with deeper cooling and high dynamic range, this portable RAMAN spectrometer delivers an improved signal to noise ratio for up to 30 minutes of integration time, making it possible to measure weak RAMAN signals.
Contact your VWR Sales Representative for more information, to request a quote or set up a demo.
Certificaciones: CDRH 1040.10; USP Chapter 1120-Raman Systems.
Información para pedidos: The i-RAMAN Plus Fiber Optic RAMAN System comes standard with a 12 month warranty and a lab grade fiber optic RAMAN Probe BAC102, one pair Laser Safety Goggles, Windows based operating software BWSpec and a trial version of BWIQ Quantitative RAMAN Analysis Software.