The Grant Optima™ TXF200 series of digitally controlled refrigerated circulating baths provides outstanding performance for routine and sophisticated applications requiring accurate temperature control in the range of -47 to 100°C
The TXF200 heating circulator incorporates Intelligent Control Optimization™ (ICO) for adaptive intelligent PID temperature control and includes a powerful integral pump (max. flow 23 L/min), making the TXF200 refrigerated systems suitable both for immersing samples and circulating temperature controlled fluid to external devices. The temperature range and heater power are automatically limited according to the liquid type selected. Combine the versatile Grant Optima™ digital immersion thermostat TXF200 with a choice of four Grant refrigeration units. Choose from a 5L or 20L tank and from a range of cooling capacities.
The Grant Optima™ TXF200 is a high-performance, programmable digital heating circulator and features a full color QVGA TFT screen displaying actual and set temperature and clear status icons. Intuitive screen icons and menus allow fast set-up of multiple parameters and provides precision control for both routine and sophisticated temperature-controlled applications in the range of -50 to 200ºC.
The Grant Optima™ TXF200 has storage for 10 programs of up to 100 segments and a choice of five languages; English, French, German, Spanish and Italian. The USB interface allows connection to PC/laptop for programming or data logging. It can be used with Labwise™ software, program to set all parameters, including programs, via the PC. Labwise™ also features real-time status windows, a graphic display of the controller's performance as a program is run, and allows data to be logged for future recall and analysis.
External circulation. Socket for external probe. Offset adjustment. Adjustable overtemperature cut out.
The series of refrigerated systems combine the powerful and versatile Grant Optima™ digital heating circulator TXF200 with a choice of four rugged stainless steel refrigeration units with different facilities and working volumes.
Certificaciones: CE Marked.
Información para pedidos: The refrigeration unit can be switched off independently of the thermostat to allow heating only applications, except for the R1 unit without relay control. All the refrigeration base units can be used up to a maximum temperature of 100°C.