- Antibody Type:Primary
- Antigen Name:kinesin family member 5B
- Antigen Symbol:KIF5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:P33176
- Antigen Synonyms:KIF5B
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:110 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store KIF5B antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human KIF5B.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10100-940
- Supplier no.:27-974
Kinesin is the founding member of a superfamily of microtubule based motor proteins that perform force-generating tasks such as organelle transport and chromosome segregation. Kinesin consists of heavy and light chains both of which have been documented to bind a variety of potential linker or cargo proteins. KIF5B is an isoform of kinesin heavy chain.
KIF5B antibody can be used for detection of KIF5B by ELISA at 1:62500. KIF5B antibody can be used for detection of KIF5B by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: KIF5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human