PHCbi's VIP® Series ultra-low temperature upright freezers are designed for safe, reliable storage of the most critical biologicals.
- Reliable
- Maximum storage
- Minimal footprint
- VIP Plus insulation composite
- Uniform preservation
- Microprocessor controls and alarms
- Five-year manufacturer's parts and labor warranty
Energy efficiency in the VIP® Series PHCbi freezer is achieved through balanced temperature performance and power management.
The integrated, microprocessor-based controller and LED display include a comprehensive set point, alarms, monitoring, and diagnostic and communication features. Ideal for storing DNA, RNA, and other biological samples, this space-saving cabinet design offers reduced wall thickness for maximum interior volume. The insulated and stainless steel framed doors with a gasket seal firmly against the cabinet to help minimize change in interior temperatures during routine door openings. The inner door latches feature an ergonomic design for one-handed operation.
The advanced combination of cabinet design, electronics, refrigeration, and critical components are engineered for enhanced security, performance and reliability. In addition, the VIP® ECO ultra-low temperature freezer operates on natural refrigerants.
Información para pedidos: All units are supplied with a five-year manufacturer's parts and labor warranty.