BRAND® 50 µl Pipette Tips, BrandTech®
Supplier: BrandTech
732106 732206 732126 732006
10034-000PK 229.22 USD
10034-000 10034-020 10034-130 10033-988
BRAND® 50 µl Pipette Tips, BrandTech®
Pipet Tips
BRAND 50 µl pipette tips are a longer style (50 mm) ultra-micro/crystal tip for use on many pipettes with a nominal/maximum volume less than 20 µl.
  • Large volume, extended length (50 mm) ultramicro/"crystal" tips
  • Fits most major brands of pipettes
  • Suitable for reverse pipetting
All tips are produced under cleanroom conditions. Bulk tips are packed in resealable bags. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.
Extended volume allows reverse pipetting of viscous or volatile liquids for greater accuracy. Graduations at 2.5, 10, 25 and 50 µl allow easy referencing of pipetted volume.
Certificaciones: CE-IVD.
Envase: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.