- Universal or flask clamp platform options
- Designed for heavy loads and continuous duty
- Safe for use in temperature-controlled environments
- Ideal for liquid cultures and general mixing
- 1-year warranty
The Orbit 1900 Shaker is a large unit designed for continuous duty and heavy loads. Its solid construction and counter-balanced drive system provide vibration-free operation. Operation of the shaker can be timed or continuous, while the speed is variable. Both time and speed are set digitally. Setting parameters is easy. Values are shown of the large 3-digit display.
Two platforms are available to accommodate a variety of different sizes of sample containers on the Orbit 1900 Shaker. The universal platform features padded bars that easily adjust to fit various flasks and bottles as well as boxes, racks, and odd-shaped containers. Flasks are accommodated on the flask platform through the use of clamps (sold separately). Custom manufactured platform/clamps are available. Suitable for use in an incubator or cold room, the Orbit 1900 Shaker fits easily inside Labnet’s High Performance 611 Series Incubators.