Airclean Systems AC600 Series Ductless Economy Workstations
Supplier: Airclean Systems
AC632A AC648A AC632TA
490005-104EA 10130.48 USD
490005-104 490005-126 490005-120
Airclean Systems AC600 Series Ductless Economy Workstations
Fume Hoods and Enclosures PCR Workstation Enclosures
  • AirSafe Automatic Safety Controller
  • Bonded carbon filtration
  • Integral base with deep spill lip
  • No ducting required
  • Clear polycarbonate shell for 360 visibility
  • Extremely quiet operation <49 dB(A)
  • Filter and electronics are located above the workspace, eliminating the chance of a chemical spill entering sensitive areas
  • Brushless, sparkless blower is post-filter
  • Deep base effectively contains accidental spills
  • Low airflow alarm warns the user both visually and audibly
  • Filter safety lockout to ensure correct filter replacement
  • Constant monitoring of airflow and filter condition
AC600 Series ductless chemical workstations are designed to provide a low cost solution for protection of the operator and environment from toxic vapors, gases, fumes, and particulate. Workstations feature the use of molded thermoplastics configured for a variety of common applications.