VWR® Storage Bottles, PP
Supplier: VWR, part of Avantor
VWR®
175-2301-VWR
89233-060EA 35.35 USD
89233-060
VWR® Storage Bottles, PP
Bottles Plastic Bottles
VWR® storage bottles are made from USP Class VI and FDA compliant resin.
  • Durable resin construction
  • Molded-in graduation marks
  • Leakproof cap
  • Polypropylene bottles are autoclavable
Storage bottles are useful for storing various acids, bases, and organic solutions.