Anti-KI67 Antibody
This antibody reacts with a nuclear antigen present in proliferating human cells. It can be used to evaluate the Ki-67 labeling index in various tumors.
Type:
Antigen: KI67
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
- Catalog No:
- 95029-736
- 99991-248
- 95025-066
- Antigen Symbol:
- KI67
- KI67
- KI67
- Antigen Name:
- Marker of proliferation Ki-67
- Marker of proliferation Ki-67
- Marker of proliferation Ki-67
- Clonality:
- Polyclonal
- Polyclonal
- Concentration:
- Ready to use
- Concentrated
- Concentrated
- SizeSupplier no.
Specifications
Antigen SymbolKI67Antigen NameMarker of proliferation Ki-67Environmentally PreferableClonalityReactivityHostIsotypeConcentrationReady to useSpecifications
Antigen SymbolKI67Antigen NameMarker of proliferation Ki-67Environmentally PreferableClonalityPolyclonalReactivityHostIsotypeConcentrationConcentratedSpecifications
Antigen SymbolKI67Antigen NameMarker of proliferation Ki-67Environmentally PreferableClonalityPolyclonalReactivityHostIsotypeConcentrationConcentrated