Omniprobe In-Situ Probe Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75960-03
102681-278PK 1019.23 USD
102681-278
Omniprobe In-Situ Probe Tips, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Custom tungsten tip with a stainless steel shank, for use with AutoProbe™ 300, in-situ probe tip exchange systems and Short-Cut™. Tip radius is 0.5 µm with 8 -10° taper angle. This tip is also Short-Cut™ compatible.