Tissue-Tek® V.I.P.™ Processing/Embedding Media, Electron Microscopy Sciences
62580-08 62580-01
102094-110CS 163.38 USD
102094-110 100498-172
Tissue-Tek® V.I.P.™ Processing/Embedding Media, Electron Microscopy Sciences
Tissue-Tek® V.I.P.™ paraffin is high-quality paraffin for processing and embedding.
  • Unique composition allows reduced compression sections as thin as 2 µm
  • Low melting point (56 °C) protects samples from heat damage
It leaves no plasticizer residue to clog paraffin lines in tissue processors and embedding centers.
Información de suministro: Supplied in small pellets for rapid melting in tissue processors/embedding centers.