EMS-820 Precision Pulsed Microwave Oven, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
97000
97100
100492-518EA
12770.98
USD
100492-518
100492-528
EMS-820 Precision Pulsed Microwave Oven, Electron Microscopy Sciences
The oven features a very well defined adjustable effect, an accurate temperature control, a well ventilated chamber, and three different timing modes for running the unit.
- Easy operation and maintenance with accurate temperature control
- Versatile with 3 different timer modes
- Built in air pump for mixing
- Manifold for up to 5 mixing tubes
- Glass fiber reinforced silicone floor
- Built in reflector for even distribution of microwaves
Temperature control not only prevents the tissue in the chamber from becoming denatured (by high temperature) but it also assists in pulsing the microwave effect in small precise portions. The temperature control has a direct effect on the magnetron (the microwave generator) as it allows the unit to switch on and off in the most efficient way at the preset effect level.
The set effect is adjustable at any time either before or during a run. This aids in the balance of the microwave load and the content of the microwave absorber. It should be noted that the penetration rate and process times are influenced by the presence of microwave radiation in the chamber and for this reason it is imperative that the effect rate be adjustable at any given time during a run.
Inside the chamber there is a manifold which allows to achieve bubble mixing in from 1 to 5 containers at once. There is a built in air pump that facilitates the mixing, avoiding temperature gradients in the mixing process. These adjustable bubble mixers make the EMS-820 ideal for decalcification. Oven is so versatile that have the option to run it in three different modes depending on application. The first mode has 3 optional presets programmed by the operator: 5 seconds-99 minutes with or without turntable. The second is timed run up to 99 minutes. And the third is continuous run for an indefinite time.
The oven chamber is made entirely of stainless steel and it is very easy to clean. It is extremely resistant to all staining solutions that are usually employed in histology and like fields. The floor of the unit, which covers the rotator cavity, is a glass fiber reinforced silicone plate which serves as a thermal insulator and will not absorb microwaves. The plate is totally chemical resistant. The unit has a built in reflector inside the chamber which evenly distributes the microwaves. A rotator is built into the unit, as well, to prevent "Hot-Spots" from forming even for very small volumes (e.g.; reagent drops on glass slides).
The EMS-820 is designed for easy operation and maintenance. All of the controls can be set with one hand and are easily accessible allowing for changes at any time. An adjustable thermal probe encased in a white "PTFE" holder (a microwave transparent material) comes with the unit. The probe can be set at different heights to accommodate all different types of containers and volumes of liquid. Attached to the probe is an adaptor to hold one of the mixing tubes; this is important because the liquid which is regulated by the thermostat needs to be mixed well to avoid temperature gradients. The probe automatically measures the existing temperature and displays it.
New Product
Limited Time Sale
Select items below are currently on sale
Enter Certificate Details
Inquire for Price
Learn more
About VWR
Item(s) Added to Cart