Double End Pestle, Electron Microscopy Sciences
64786-02
100498-874EA 101.97 USD
100498-874
Double End Pestle, Electron Microscopy Sciences
This pestle is mounted on a stainless steel shaft, 3.2mm (1/8") in diameter and it may be hand held or mounted in a motorized chuck. Double ended pestle. One end fits the 1.5 ml microcentrifuge tube and the other end fits the 0.5 ml tube. Overall length is 152mm (6").