Dissecting Boards, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63307-30 63307-50 63307-20
100500-996BX 170.96 USD
100500-996 100498-398 100498-396
Dissecting Boards, Electron Microscopy Sciences
Boards are available in three sizes.
Made of 5/8" thick high-density polyethylene. A white background with X, Y scales in centimeters offers a quick orientation and sizing of the specimens. It is chemical and stain resistant and comes with a spill control border and rubber feet to prevent spillage and sliding. To secure the specimen to the board, styrene-capped pins are available (optional).
Boards are available in three sizes.
Board A L x W x H: 28 x 22.8 x 1.6 cm (11 x 9 x 5/8")
Board B L x W x H: 33 x 28 x 1.6 cm (13 x 11 x 5/8")
Board C L x W x H: 58.4 x 40.6 x 1.6 cm (23 x 16 x 5/8")