SP Bel-Art Large Polyethylene Policeman, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37739-0002
13197-254BG 286.87 USD
13197-254
SP Bel-Art Large Polyethylene Policeman, Bel-Art Products, a part of SP
Spatulas
The handle of this policeman contains a rigid 6 mm (¼") steel rod enclosed in polyethylene.
  • Rigid design
  • Polyethylene coating over stainless steel core
  • Efficient
The tapered flattened end is 25 mm (1×1") for efficient scraping and transfer of materials.
Envase: Blister packed.