Puerto Rico
Order Entry
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
 
undefined
Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 99990-894
  • 95024-340
  • 99990-292
  • Antigen Symbol:
  • CD34
  • CD34
  • CD34
  • Antigen Name:
  • CD34 molecule
  • CD34 molecule
  • CD34 molecule
  • Antigen Synonyms:
  • CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
  • CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
  • CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Clone:
  • QBEND/10
  • QBEND/10
  • QBEND/10
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Concentration:
  • Ready to use
  • Concentrated
  • Concentrated

 

 

  •  
     
    Size
    Supplier no.
     
     
  • 95024-340
    MOB098-05
    500 µL
    MOB 098-05
       
       
    Each
    Each - $494.07
    $494.07
    Each
       $494.07

     
     
     
    Specifications
    Antigen Symbol
    CD34
    Antigen Name
    CD34 molecule
    Environmentally Preferable
    Antigen Synonyms
    CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
    Clonality
    Monoclonal
    Clone
    QBEND/10
    Reactivity
    Host
    Mouse
    Isotype
    Concentration
    Concentrated
  • 99990-292
    MOB098
    1 mL
    MOB 098
       
       
    Each (1ml)
    Each (1ml) - $817.22
    $817.22
    Each (1ml)
       $817.22

     
     
     
    Specifications
    Antigen Symbol
    CD34
    Antigen Name
    CD34 molecule
    Environmentally Preferable
    Antigen Synonyms
    CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
    Clonality
    Monoclonal
    Clone
    QBEND/10
    Reactivity
    Host
    Mouse
    Isotype
    Concentration
    Concentrated
  • 99990-894
    PDM050
    6 mL
    PDM 050
       
       
    Each (6ml)
    Each (6ml) - $536.60
    $536.60
    Each (6ml)
       $536.60

     
     
     
    Specifications
    Antigen Symbol
    CD34
    Antigen Name
    CD34 molecule
    Environmentally Preferable
    Antigen Synonyms
    CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
    Clonality
    Monoclonal
    Clone
    QBEND/10
    Reactivity
    Host
    Mouse
    Isotype
    Concentration
    Ready to use