Pumice stone, granular for elementary analysis, Supelco®

Supplier: MilliporeSigma

1.02441.1000 1.02441.1000
EM1.02441.1000CS 2336 USD
CA1.02441.1000 EM1.02441.1000
Pumice stone, granular for elementary analysis, Supelco®
Pumice stone
MDL Number: MFCD00148147
CAS Number: 1332-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR