Thermometers, Fractional Degree, Ace Glass Incorporated
Supplier: Ace Glass
8360-04 8360-01 8360-06
89084-778EA 232.67 USD
89084-778 89084-776 89084-774
Thermometers, Fractional Degree, Ace Glass Incorporated
Thermometers Analog Thermometers
These thermometers feature fractional degree divisions
They are red spirit filled with white backed glass and permanent fused markings. Meets ANSI/SAMA Z236.1 – 1983 standards.